Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Momp Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | Chlamydia Momp |
---|---|
Concentration | 4.0 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunofluorescence, Immunocytochemistry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017688
![]() |
Novus Biologicals
NB100650540.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10065054
![]() |
Novus Biologicals
NB10065054 |
0.5 mL |
Each for $501.50
|
|
|||||
Description
Chlamydia Momp Polyclonal specifically detects Chlamydia Momp in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Chlamydia Momp | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline with 0.09% Sodium Azide | |
OMPA | |
IgG | |
IgG purified | |
Recognizes L2 MOMP and purified EBs (all serovars, A-K, L1-L3). |
4.0 mg/mL | |
Immunofluorescence, Immunocytochemistry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Goat | |
Bacteria | |
Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2 | |
Purified MOMP (major outer membrane protein) of Chlamydia trachomatis strain L2. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title