Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Trachomatis Lps Antibody (1312/236), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1066402A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066402A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066402A70 Supplier Novus Biologicals Supplier No. NB10066402AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chlamydia Trachomatis Lps Monoclonal specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Chlamydia Trachomatis Lps
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1312/236
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Native
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity In a simple ELISA this is reactive with 15 serovars of C. trachomatis. The epitope has been shown by Western blotting to be present in LPS.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.