Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Trachomatis Lps Antibody (1312/236), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB211710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211710 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211710 Supplier Novus Biologicals Supplier No. NB10066402JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chlamydia Trachomatis Lps Monoclonal antibody specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen Chlamydia Trachomatis Lps
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1312/236
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.