Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Trachomatis Lps Antibody (1312/284), Novus Biologicals™
SDP

Catalog No. NBP128820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128820 0.1 mg
NB018472 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP128820 Supplier Novus Biologicals Supplier No. NBP128820
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chlamydia Trachomatis Lps Monoclonal specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Chlamydia Trachomatis Lps
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1312/284
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Host Species Mouse
Immunogen Chlamydia trachomatis L2
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CHLAMYDIA TRACHOMATIS LPS
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.