Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Trachomatis Momp Antibody (1297/143), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB166403AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166403AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB166403AF Supplier Novus Biologicals Supplier No. NB10066403AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chlamydia Trachomatis Momp Monoclonal specifically detects Chlamydia Trachomatis Momp in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Chlamydia Trachomatis Momp
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1297/143
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2
Gene Symbols O175_03780
Host Species Mouse
Immunogen Native type E MOMP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with MOMP from C. trachomatis.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.