Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Trachomatis Momp Antibody (1297/143), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1066403A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066403A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066403A32 Supplier Novus Biologicals Supplier No. NB10066403AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chlamydia Trachomatis Momp Monoclonal specifically detects Chlamydia Trachomatis Momp in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Chlamydia Trachomatis Momp
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1297/143
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2
Host Species Mouse
Immunogen Native type E MOMP
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Reacts with MOMP from C. trachomatis.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.