Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Trachomatis Momp Antibody (1297/143) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017770 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017770 0.025 mg
NB10066403 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017770 Supplier Novus Biologicals Supplier No. NB100664030.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chlamydia Trachomatis Momp Monoclonal specifically detects Chlamydia Trachomatis Momp in Bacteria, Rhesus Macaque samples. It is validated for Dot Blot, ELISA, Western Blot.

Specifications

Antigen Chlamydia Trachomatis Momp
Applications Dot Blot, ELISA, Western Blot
Classification Monoclonal
Clone 1297/143
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P17451
Gene Alias Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2
Gene Symbols O175_03780
Host Species Mouse
Immunogen Native type E MOMP
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with MOMP from C. trachomatis.
Target Species Bacteria, Rhesus Macaque
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.