Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Trachomatis Momp Antibody (1297/143) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Chlamydia Trachomatis Momp |
---|---|
Clone | 1297/143 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017770
![]() |
Novus Biologicals
NB100664030.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10066403
![]() |
Novus Biologicals
NB10066403 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Chlamydia Trachomatis Momp Monoclonal specifically detects Chlamydia Trachomatis Momp in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Chlamydia Trachomatis Momp | |
1.0 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2 | |
Native type E MOMP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1297/143 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
P17451 | |
O175_03780 | |
IgG2a | |
Protein A purified | |
Reacts with MOMP from C. trachomatis. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title