Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloramphenicol Antibody - BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Chloramphenicol |
---|---|
Concentration | 5.0 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017476
![]() |
Novus Biologicals
NB100624570.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10062457
![]() |
Novus Biologicals
NB10062457 |
0.5 mL |
Each for $499.50
|
|
|||||
Description
Chloramphenicol Polyclonal specifically detects Chloramphenicol in Non-species specific samples. It is validated for ELISA.Specifications
Chloramphenicol | |
ELISA 1:100-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.09% Sodium Azide | |
IgG | |
IgG purified | |
This product recognises chloramphenicol, a broad-spectrum antimicrobial that inhibits protein synthesis. |
5.0 mg/mL | |
ELISA | |
Unconjugated | |
Sheep | |
Non-species specific | |
Chloramphenicol base conjugated via its amino group to KLH using phenylenediisothiocyanate | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title