Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroform, Stabilized, Reagent, ACS, 99.8%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
Description
Spectrum™ Chemical Chloroform, Stabilized, Reagent, ACS , is a colorless, sweet smelling organic compound comes in the form of a dense liquid and is a trihalomethane. It is also commonly used as laboratory solvent because it is relatively unreactive, conveniently volatile and is miscible with most organic compounds. Although it uses as a reagent are limited, it is used in organic synthesis in order to produce dichlorocarbene by reacting with sodium hydroxide. As an ACS grade Reagent, Spectrum Chemical manufactured chloroform is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 100 mL |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 0.001% |
| UN Number | UN1888 |
| Percent Purity | 99.8% |
| Grade | ACS |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction