Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP1B Antibody, Novus Biologicals™
SDP

Catalog No. NBP155512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155512 Supplier Novus Biologicals Supplier No. NBP155512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHMP1B Polyclonal specifically detects CHMP1B in Human samples. It is validated for Western Blot.

Specifications

Antigen CHMP1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q7LBR1
Gene Alias C18orf2C10orf2, charged multivesicular body protein 1b, CHMP1.5C18-ORF2, CHMP1b, chromatin modifying protein 1B, Chromatin-modifying protein 1b, hVps46-2, vacuolar protein sorting 46-2, Vacuolar protein sorting-associated protein 46-2, Vps46-2, Vps46B
Gene Symbols CHMP1B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CHMP1B(chromatin modifying protein 1B) Antibody(against the N terminal of CHMP1B. Peptide sequence KIKKAIQKGNMEVARIHAENAIRQKNQAVNFLRMSARVDAVAARVQTAVT.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57132
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.