Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP1B Antibody, Novus Biologicals™
SDP

Catalog No. NB397016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397016 25 μL
NBP237986 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397016 Supplier Novus Biologicals Supplier No. NBP23798625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHMP1B Polyclonal specifically detects CHMP1B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CHMP1B
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q7LBR1
Gene Alias C18orf2C10orf2, charged multivesicular body protein 1b, CHMP1.5C18-ORF2, CHMP1b, chromatin modifying protein 1B, Chromatin-modifying protein 1b, hVps46-2, vacuolar protein sorting 46-2, Vacuolar protein sorting-associated protein 46-2, Vps46-2, Vps46B
Gene Symbols CHMP1B
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: QNQVDMLLQEMADEAGLDLNMELPQGQTGSVGTSVASAEQDELSQRLARLRDQV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57132
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.