Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP4A Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB145609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB145609 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB145609 Supplier Novus Biologicals Supplier No. NBP306005AF647

Rabbit Polyclonal Antibody

CHMP4A Polyclonal specifically detects CHMP4A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CHMP4A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C14orf123, charged multivesicular body protein 4a, CHMP4, CHMP4a, CHMP4B, chromatin modifying protein 4A, Chromatin-modifying protein 4a, chromosome 14 open reading frame 123, FLJ61658, hSnf-1, HSPC134, hVps32-1, MGC142093, MGC142095, SHAX2, SNF7, SNF7 homolog associated with Alix-2, Snf7 homologue associated with Alix 2, SNF7-1, Vacuolar protein sorting-associated protein 32-1, VPS32-1, VPS32A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CHMP4A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29082
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.