Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP4C Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB332186 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332186 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332186 Supplier Novus Biologicals Supplier No. NBP297351MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHMP4C Polyclonal antibody specifically detects CHMP4C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen CHMP4C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias charged multivesicular body protein 4c, CHMP4c, chromatin modifying protein 4C, Chromatin-modifying protein 4c, hSnf7-3, hVps32-3, MGC22825, Shax3, SNF7 homolog associated with Alix 3, Snf7 homologue associated with Alix 3, SNF7-3, Vacuolar protein sorting-associated protein 32-3, Vps32-3, VPS32C
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CHMP4C fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92421
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.