Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHMP4C Antibody, Novus Biologicals™
SDP

Catalog No. NB034807 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB034807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB034807 Supplier Novus Biologicals Supplier No. NBP287185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHMP4C Polyclonal specifically detects CHMP4C in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CHMP4C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias charged multivesicular body protein 4c, CHMP4c, chromatin modifying protein 4C, Chromatin-modifying protein 4c, hSnf7-3, hVps32-3, MGC22825, Shax3, SNF7 homolog associated with Alix 3, Snf7 homologue associated with Alix 3, SNF7-3, Vacuolar protein sorting-associated protein 32-3, Vps32-3, VPS32C
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse CHMP4C
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92421
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.