Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHN 1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB080548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080548 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080548 Supplier Novus Biologicals Supplier No. NBP296984C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHN 1 Polyclonal specifically detects CHN 1 in Human samples. It is validated for ELISA.

Specifications

Antigen CHN 1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias a2-chimaerin, A-chimaerin, alpha-chimerin, ARHGAP2RhoGAP2, chimerin (chimaerin) 1, CHNDuane retraction syndrome 2, DURS2, NC, N-chimaerin, N-chimerin, Rho GTPase-activating protein 2, RHOGAP2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CHN 1 (NP_001813.1; Ala2-Phe459).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1123
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.