Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHN 1 Antibody (OTI2G6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N270411J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270411J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270411J646 Supplier Novus Biologicals Supplier No. NBP270411JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHN 1 Monoclonal specifically detects CHN 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CHN 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2G6
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias a2-chimaerin, A-chimaerin, alpha-chimerin, ARHGAP2RhoGAP2, chimerin (chimaerin) 1, CHNDuane retraction syndrome 2, DURS2, NC, N-chimaerin, N-chimerin, Rho GTPase-activating protein 2, RHOGAP2
Gene Symbols CHN1
Host Species Mouse
Immunogen Full length recombinant protein of human CHN 1 (NP_001813) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1123.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.