Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholera Haemolysin Antibody (2024/251), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB162460AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162460AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162460AH Supplier Novus Biologicals Supplier No. NB10062460AF647

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Cholera Haemolysin Monoclonal specifically detects Cholera Haemolysin in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Cholera Haemolysin
Applications ELISA
Classification Monoclonal
Clone 2024/251
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HlyA, VCA0219
Host Species Mouse
Immunogen Native organisms
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62460 is specific for Vibrio cholerae haemolysin (HlyA). HlyA is an extracellular matrix damaging protein with a molecular weight of 65 kDa that exists a water-soluble monomer and an oligomeric amphipathic protein. It belongs to a class of proteins known as pore-forming toxins for their ability to punch holes in plasma membranes.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.