Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cholera Toxin Alpha Mouse anti-Bacteria, Clone: 2/166, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100624610.025MG
Description
Cholera Toxin Alpha Monoclonal specifically detects Cholera Toxin Alpha in Bacteria samples. It is validated for ELISA.Specifications
Cholera Toxin Alpha | |
Monoclonal | |
5.0 mg/mL | |
ELISA 1:100-1:2000 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
NB100-62461 is specific for the alpha subunit of Cholera toxin. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM |
ELISA | |
2/166 | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Vibrio cholerae extract | |
0.025 mg | |
Primary | |
Bacteria | |
Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction