Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cholera Toxin Alpha Mouse anti-Bacteria, Clone: 2/166, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00
Specifications
| Antigen | Cholera Toxin Alpha |
|---|---|
| Clone | 2/166 |
| Concentration | 5.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
Description
Cholera Toxin Alpha Monoclonal specifically detects Cholera Toxin Alpha in Bacteria samples. It is validated for ELISA.Specifications
| Cholera Toxin Alpha | |
| 5.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| IgM | |
| Ammonium sulfate precipitation | |
| NB100-62461 is specific for the alpha subunit of Cholera toxin. |
| 2/166 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Vibrio cholerae extract | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title