Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholera Toxin Beta Antibody (23043), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10064675V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064675V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064675V Supplier Novus Biologicals Supplier No. NB10064675V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cholera Toxin Beta Monoclonal specifically detects Cholera Toxin Beta in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Cholera Toxin Beta
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 23043
Conjugate DyLight 405
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cholera enterotoxin B chain, Cholera enterotoxin beta chain, Cholera enterotoxin gamma chain, Cholera enterotoxin subunit B, Cholera toxin B protein, Choleragenoid, CTX B, CTXB, TOX B, toxB, VC1456
Host Species Mouse
Immunogen Vibrio cholerae
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64675 detects the labile enterotoxin normally associated with V. cholera and reacts with an epitope on the B chain. No reactivity was observed with other species tested including various E. coli serotypes.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.