Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase/ChAT Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB10098693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10098693 0.5 mg
NB017939 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10098693 Supplier Novus Biologicals Supplier No. NB10098693

Sheep Polyclonal Antibody

Choline Acetyltransferase/ChAT Polyclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen Choline Acetyltransferase/ChAT
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml
Formulation Lyophilized from PBS with No Preservative
Gene Accession No. Q03059, P32738
Gene Alias acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6
Gene Symbols CHAT
Host Species Sheep
Immunogen A synthetic peptide from mouse Choline Acetyltransferase/ChAT conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and human.
Purification Method Ammonium sulfate precipitation
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1103
Test Specificity ChAT.
Reconstitution Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.