Learn More
Chondroitin sulfate, 90+%
Description
Chondroitin sulfate is used in a wide variety of areas of research such as bioengineering. Chondroitin sulfate may be used as a biomaterial copolymer or surface derivatization reagent in the development of drug delivery vehicles, tissue engineering devices and bioscaffolds. Chondroitin sulfate may be used for the development of biocompatible structures such as hydrogels, sponges, biofilms, microspheres and micelles. Used in the treatment of coronary atherosclerosis, myocardial ischemia, myocardial infraction. Chondroitin sulfate is a sulfated (in different positions) glycosaminoglycan (GAG), composed of repeating and alternating disaccharide units of D-glucuronic acid and N-acetylgalactosamine. Depending on the position of sulfate, Chondroitin sulfate is named as chondroitin sulfate A (sulfate on carbon 4 of the N-acetylgalactosamine (GalNAc) sugar, C (carbon 6 of the GalNAc sugar), D (carbon 2 of the glucuronic acid and 6 of the GalNAc sugar) and E (carbons 4 and 6 of the GalNAc sugar). The CAS # given is the representation of core component of this product.
Chondroitin sulfate is used in a wide variety of areas of research such as bioengineering. Chondroitin sulfate may be used as a biomaterial copolymer or surface derivatization reagent in the development of drug delivery vehicles, tissue engineering devices and bioscaffolds. Chondroitin sulfate may be used for the development of biocompatible structures such as hydrogels, sponges, biofilms, microspheres and micelles. Used in the treatment of coronary atherosclerosis, myocardial ischemia, myocardial infraction.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Chemical Name or Material | Chondroitin sulfate |
| Color | White |
| Quantity | 25 g |
| MDL Number | MFCD00146419 |
| Merck Index | 14,2214 |
| Synonym | chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid |
| Solubility Information | Soluble in water. |
| InChI Key | KXKPYJOVDUMHGS-OSRGNVMNSA-N |
| SMILES | CC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O |
| IUPAC Name | (2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.