Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chondroitin Sulfate (NG2) Mouse anti-Human, PE, Clone: 7.1, BD Phrmingen™
Mouse Monoclonal Antibody
$128.00 - $312.00
Specifications
Antigen | Chondroitin Sulfate (NG2) |
---|---|
Clone | 7.1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Chondroitin Sulfate (NG2) Monoclonal antibody specifically detects Chondroitin Sulfate (NG2) in Human samples. It is validated for Flow cytometrySpecifications
Chondroitin Sulfate (NG2) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
1464 | |
IgG1 κ | |
Affinity Chromatography |
7.1 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
CSPG4; HMW-MAA; MCSP; MCSPG; NG2 | |
Human BM stromal cell line established by infection with SV-40 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title