Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondrolectin Antibody (396305), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB313374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313374 Supplier Novus Biologicals Supplier No. FAB2576PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chondrolectin Monoclonal antibody specifically detects Chondrolectin in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Chondrolectin
Applications Flow Cytometry
Classification Monoclonal
Clone 396305
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias C21orf68, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chondrolectin, Arg22-Asn216, Accession # Q9H9P2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 140578
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.