Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHORDC1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP178304A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP178304A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP178304A70 Supplier Novus Biologicals Supplier No. NBP178304AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHORDC1 Polyclonal specifically detects CHORDC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CHORDC1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CHORD domain-containing protein 1, CHORD-containing protein 1, CHP1CHP-1, cysteine and histidine-rich domain (CHORD) containing 1, cysteine and histidine-rich domain (CHORD)-containing 1, cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1, cysteine and histidine-rich domain-containing protein 1, FLJ37289, Protein morgana
Gene Symbols CHORDC1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 26973
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.