Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chordin-like 2/CHRDL2 Antibody (272905), HRP, Novus Biologicals™
SDP

Catalog No. NB365119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365119 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365119 Supplier Novus Biologicals Supplier No. FAB24481H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chordin-like 2/CHRDL2 Monoclonal antibody specifically detects Chordin-like 2/CHRDL2 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Chordin-like 2/CHRDL2
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 272905
Conjugate HRP
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias chordin-like 2, chordin-like protein 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chordin-like 2/CHRDL2, Arg28-Thr451, Accession # NP_056239
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25884
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.