Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (77F12), Biotin, Novus Biologicals™
SDP

Catalog No. NB001743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001743 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001743 Supplier Novus Biologicals Supplier No. NB10073002B200UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Sandwich ELISA.

Specifications

Antigen Chorionic Gonadotropin alpha Chain (hCG alpha)
Applications Sandwich ELISA
Classification Monoclonal
Clone 77F12
Conjugate Biotin
Dilution ELISA
Formulation PBS
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen This specifically recognizes HCG; it reacts mainly with the alpha-subunit. It is cross-reacting with the whole molecule of HCG, hLH, hTSH and hFSH. Cross-reactivity with free beta-subunit of HCG was not detected.
Quantity 200 μL
Gene ID (Entrez) 1081
Test Specificity This specifically recognizes HCG; it reacts mainly with the alpha-subunit. It is cross-reacting with the whole molecule of HCG, hLH, hTSH and hFSH. Cross-reactivity with free beta-subunit of HCG was not detected.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.