Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (INN-hFSH-132), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB162251FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162251FR 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162251FR Supplier Novus Biologicals Supplier No. NB10062251FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Chorionic Gonadotropin alpha Chain (hCG alpha)
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone INN-hFSH-132
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen Human Follicle Stimulating Hormone (FSH)
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Test Specificity A high affinity antibody recognising the alpha 4 epitope of the alpha-subunit of Human Chorionic Gonadotrophin (hCG), hLH, hFSH, hTSH and free alpha-subunit. The recognised epitope comprises amino-acids 15-22 of the human sequence. This antibody will not cross-react with glycoprotein hormones of various animal species.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.