Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (INN-hFSH-158), FITC
SDP

Catalog No. NB10062226F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062226F 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062226F Supplier Novus Biologicals Supplier No. NB10062226F

Mouse Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for ELISA.

Specifications

Antigen Chorionic Gonadotropin alpha Chain (hCG alpha)
Applications ELISA
Classification Monoclonal
Clone INN-hFSH-158
Conjugate FITC
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen Human FSH
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Test Specificity This antibody reacts with the alpha 5 epitope on the alpha subunit of hCG, hLH, hFSH, hISH and the free alpha subunit. The alpha 5 epitope is destroyed upon iodination.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.