Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (LHa/756) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Chorionic Gonadotropin alpha Chain (alpha HCG) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | LHa/756 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1 - 2 ug/ml |
| Formulation | PBS with No Preservative |
| Gene Alias | CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, HCG, LHA, TSHA |
| Gene Symbols | CGA |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.