Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (LHa/756), Novus Biologicals™
SDP

Catalog No. NB253394200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253394200 200 μg
NB25339420 20 μg
NB253394100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253394200 Supplier Novus Biologicals Supplier No. NBP253394200UG

Mouse Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen Chorionic Gonadotropin alpha Chain (alpha HCG)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LHa/756
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1 - 2 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, HCG, LHA, TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen Recombinant full-length hLH alpha protein
Molecular Weight of Antigen 13 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.