Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (SPM552), Novus Biologicals™
SDP

Catalog No. NBP232971A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232971A 0.1 mg
NBP232971F 0.02 mg
NBP232971B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP232971A Supplier Novus Biologicals Supplier No. NBP2329710.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Chorionic Gonadotropin alpha Chain (HCG alpha)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM552
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P01215
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Gene Symbols CGA
Host Species Mouse
Immunogen Recombinant hCG alpha protein
Molecular Weight of Antigen 13 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Test Specificity This MAb reacts with a protein of ∼13kDa, identified as alpha sub-unit of HCG. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the ? and ? subunits. The subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.