Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Rabbit anti-Human, Clone: HCGa/2728R, Novus Biologicals™
SDP

Catalog No. NB177645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177645 20 μg
NB177644 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177645 Supplier Novus Biologicals Supplier No. NBP31384420UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Chorionic Gonadotropin alpha Chain (hCG alpha)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HCGa/2728R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P01215
Gene Alias CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Host Species Rabbit
Immunogen Recombinant fragment (around aa1-100) of human Chorionic Gonadotropin alpha Chain (hCG alpha) protein (exact sequence is proprietary) (Uniprot: P01215)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1081
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.