Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin beta Chain (hCG beta) Antibody (HCGb/54+ HCGb/459), Novus Biologicals™
SDP

Catalog No. NBP234238B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234238B 0.2 mg
NBP234238F 0.02 mg
NBP234238A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP234238B Supplier Novus Biologicals Supplier No. NBP2342380.2MG

Mouse Monoclonal Antibody

Chorionic Gonadotropin beta Chain (HCG beta) Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting.

Specifications

Antigen Chorionic Gonadotropin beta Chain (HCG beta)
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HCGb/54+ HCGb/459
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P01233
Gene Alias CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB
Gene Symbols CGB
Host Species Mouse
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Molecular Weight of Antigen 22 kDa
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1082
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.