Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-24) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018249 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018249 0.05 mg
NB600684 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018249 Supplier Novus Biologicals Supplier No. NB6006840.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for ELISA, Radioimmunodiffusion.

Specifications

Antigen hCG beta 4
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone INN-hCG-24
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Radioimmunodiffusion
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Gene Accession No. P01233
Gene Alias CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB
Gene Symbols CGB3
Host Species Mouse
Immunogen Human CG.
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1082
Test Specificity This is a high affinity antibody recognising the beta 4 epitope on hCG. It is also reactive with hCGbeta, hCGbetacf (fragment), hCGn (nicked), hCGbetan, CTP-hCG, CTP-hCGbeta. It may be used in combination with INN hCG2, 32 and 45. It has minor cross reactivity with hLH and hLHbeta. No cross reaction is observed with hFSH, hTSH, or the glycoprotein hormone alpha chains.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.