Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-45), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB162225AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162225AG 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162225AG Supplier Novus Biologicals Supplier No. NB10062225AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hCG holo C3 Monoclonal specifically detects hCG holo C3 in Human samples. It is validated for ELISA, Radioimmune Assays (RIA).

Specifications

Antigen hCG holo C3
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone INN-hCG-45
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGB chorionic gonadotropin, beta polypeptide, CGB3, CGB5, CGB7, CGB8, hCGB
Gene Symbols CGB3
Host Species Mouse
Immunogen Human hCG holo C3
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1082
Test Specificity This antibody reacts with hCG holo C3 recognising the c3 epitope on hCG. No subunit detection is observed.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.