Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017540 0.05 mg
NB10063167 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017540 Supplier Novus Biologicals Supplier No. NB100631670.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hCG holo C2 Monoclonal specifically detects hCG holo C2 in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen hCG holo C2
Applications ELISA, Western Blot
Classification Monoclonal
Clone INN-hCG-53
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P01233
Gene Alias choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB
Gene Symbols CGB3
Host Species Mouse
Immunogen Made to Human hCG holo C2
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1082
Test Specificity NB100-63167 recognizes epitope c2 on hCG holo C2 . It does not recognize free hCG subunits.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.