Learn More
Eisco™ Chorite Mineral Specimen
SCI_ED
Catalog No. S27575 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27575 1/Ea.
S27597 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27575 Supplier Eisco™ Supplier No. ESNG0068
Add to Cart
Add to Cart
Chlorite is a metamorphic mineral that has undergone light alteration. During metasomatism, phyllosilicate minerals may become hydrated and form chlorite.
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for lessons on metamorphism, mineral alteration, and general identification exercises
  • The greener color and near-soapy feel of chlorite distinguishes it from other phyllosilicates at a glance, and the presence of water distinguishes it chemically
Specifications
Product Type Mineral
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.05 kg