Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chp2 Antibody (CPTC-CHP2-1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB391360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391360 Supplier Novus Biologicals Supplier No. NBP308694AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chp2 Monoclonal antibody specifically detects Chp2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Chp2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-CHP2-1
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias calcineurin B homologous protein 2, HCA520, hepatocellular carcinoma antigen gene 520, Hepatocellular carcinoma-associated antigen 520
Host Species Mouse
Immunogen Recombinant human full-length Chp2 protein (Uniprot: O43745)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63928
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.