Learn More
CHREBP Antibody (2D9NB) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 3 publications
Supplier:  Novus Biologicals NBP244307
 View more versions of this product
Catalog No. NBP244307
Only null left
Add to Cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
CHREBP Monoclonal specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).
Specifications
Specifications
CHREBP
Monoclonal
1.0 mg/mL
Western Blot 1:500 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 31289120), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP)
Q9NP71-1
MLXIPL
Partial recombinant human ChREBP protein between amino acids 600-800. [Uniprot: Q9NP71]
0.1 mg
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators
51085
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b κ
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
2D9NB
Unconjugated
PBS with 0.02% Sodium Azide
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only