Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHREBP Antibody (2D9NB) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP244307SS
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
CHREBP Monoclonal specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).Specifications
CHREBP | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:500 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 31289120), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) | |
Q9NP71-1 | |
MLXIPL | |
Partial recombinant human ChREBP protein between amino acids 600-800. [Uniprot: Q9NP71] | |
0.025 mg | |
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators | |
51085 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
2D9NB | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction