Learn More
Description
Specifications
Specifications
| Antigen | CHREBP |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Immunoassay |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockout Validated |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein |
| Gene Symbols | MLXIPL |
| Host Species | Rabbit |
| Immunogen | A C-terminal synthetic peptide made to the human CHREBP protein sequence (between residues 800-852). [UniProtNo. Q9NP71, Isoform 1/Alpha] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
