Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHREBP Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB354845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354845 Supplier Novus Biologicals Supplier No. NB400135PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHREBP Polyclonal antibody specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, Gel Super Shift Assays, ChIP assay (ChIP), KnockDown

Specifications

Antigen CHREBP
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Supershift, SDS-Page, ChIP Assay, Gene Knock-Out
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Gel Super Shift Assays, SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated
Formulation PBS
Gene Alias BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to the human CHREBP protein sequence (between residues 800-852). [UniProt# Q9NP71, Isoform 1/Alpha]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51085
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.