Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Chroma-Luc™ Luciferase Reporter Vectors
Click to view available options
Product Type:
pCBG68-Basic Vector
pCBG68-Control Vector
pCBG99-Basic Vector
pCBG99-Control Vector
pCBR-Basic Vector
pCBR-Control Vector
Description
- Control Vectors That Can Generate Red and Green (Dual-Color) Luminescence in a Single Sample
- Use as the control reporter vector with Chroma-Luc™ Basic Vectors
- Engineered genes are based on click beetle luciferase
- Luciferase expression driven by SV40 promoter and enhancer
Specifications
Specifications
For Use With (Equipment) | Chroma-Glo assay system |
Promoter | SV40 |
Product Type | pCBR-Basic Vector |
Reporter Gene | Luciferase |
Content And Storage | -30°C to -10°C |
Quantity | 20 μg |
For Use With (Application) | Cloning regulatory elements of interest to produce red/green (dual color) luminescence |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction