Learn More
Promega Chroma-Luc™ Luciferase Reporter Vectors
Chroma-Glo™ Luciferase Assay System and the Chroma-Luc Vectors are used to generate red and green (dual-color) luminescence from a single sample upon addition of a single reagent.
Manufacturer:  Promega E1461
 View more versions of this product
Catalog No. PRE1461
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Control Vectors That Can Generate Red and Green (Dual-Color) Luminescence in a Single Sample
  • Use as the control reporter vector with Chroma-Luc™ Basic Vectors
  • Engineered genes are based on click beetle luciferase
  • Luciferase expression driven by SV40 promoter and enhancer
Specifications
Specifications
Cloning regulatory elements of interest to produce red/green (dual color) luminescence
SV40
Luciferase
Chroma-Glo assay system
pCBG99-Control Vector
20 μg