Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ CHROMAFLEX™ Flow Adapters
SDP

Catalog No. p-6899048 Shop All DWK Life Sciences Products
Click to view available options
Description:
Fits column ID: 1 cm
Fits column ID: 2.5 cm
Fits column ID: 4.8 cm
Diameter (English) Outer:
1/16 in.
1/8 in.

Flow adapters improve chromatogram resolution by delivering the sample and buffer directly to the top of the gel bed. This protects the gel bed from disruption during sample loading and eliminates any dead volume above the gel bed.

  • Flow adapters are recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
  • Each flow adapter is supplied with a 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 feet (1.5 meters) of FEP PTFE tubing
  • Blue lock ring is supplied with all sizes except those that fit 1.0 cm column ID
  • Use of aggressive organic solvents requires the PTFE bed supports listed as accessories and FFKM o-rings sizes 010, 116 or 222
TRUSTED_SUSTAINABILITY

Specifications

Type Fits Column: Acrylic Jacketed
Diameter (English) Outer 1/16 in.
Temperature (English) Max. 122°F
For Use With (Application) Improving Chromatogram Resolution
Includes 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 ft. (1.5 meters) of FEP PTFE tubing
Description Fits column ID: 1 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.