Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Oxide (Powder/Technical), Fisher Chemical

Catalog No. c3333 Shop All Fisherbrand Products
Click to view available options
Quantity:
3 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy

Catalog No. C333-3

Only null left
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 4000°C
Melting Point 2435°C
Color Green
pH 5 to 7
Physical Form Powder/Solid
Quantity 3 kg
UN Number UN 1463
Formula Weight 151.99
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Density 5.210g/cm³
Chemical Name or Material Chromic Oxide
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.