Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Potassium Sulfate Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical

Catalog No. c337500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle

Catalog No. C337-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrH24KO20S2
Molecular Weight (g/mol) 499.386
MDL Number MFCD00149917
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

Specifications

Melting Point 88.9°C
Color Purple
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 102.0 %
Linear Formula CrK(SO4)2·12H2O
Formula Weight 499.4
Percent Purity 98.0 to 102.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 1.8g/cm³
Chemical Name or Material Chromic Potassium Sulfate Dodecahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.