Chromium(III) nitrate nonahydrate, 98.5%, Alfa Aesar

For Use As a Catalyst, Corrosion Inhibitor, in Textile Printing

Manufacturer: Alfa Aesar 1156514

Quantity 25g

 View more versions of this product

Catalog No. AA1156514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-02-8
    Physical Form Crystalline
    Assay Percent Range 98.5%
    Molecular Formula Cr(NO3)3·9H2O
    Formula Weight 400.14 (238.01 Anhydrous)
    MDL Number MFCD00149659
    Chemical Name or Material Chromium(III) nitrate nonahydrate, nonahydrate

    Specifications

    Formula Weight 400.14 (238.01 Anhydrous)
    Quantity 25g
    Color Purple
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 236-921-1
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 98.5%
    Molecular Formula Cr(NO3)3·9H2O
    Appearance Purple crystalline
    MDL Number MFCD00149659
    CAS 7789-02-8
    Health Hazard 1 H272-H315-H319-H303 : May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May be harmful if swallowed.
    Chemical Name or Material Chromium(III) nitrate nonahydrate, nonahydrate

    H272-H315-H319-H303 : May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit